2600 Disposable N95 Particulate Respirators, Moldex®
Supplier: Moldex/Metric
2600N95
46610-306CS 628.49 USD
46610-306
2600 Disposable N95 Particulate Respirators, Moldex®
Respirators Disposable Respirators Particulate Respirators
The particulate respirators are made with Softspun® lining for added comfort and durability.
  • Stretch cloth strap feels cool and comfortable against the skin
  • Featuring a contoured nose bridge with soft foam nose flange, this respirator allows all-day comfort
  • Dura-Mesh® shell will not collapse in heat and humidity
  • Certified to have a filter efficiency of 95% or greater against particulates free of oil
The respirators have a cloth HandyStrap® with a buckle allowing the mask to hang around the neck when not in use.