VWR® Lab Wrap, Clear Plastic
Supplier: VWR International
3470
46610-056EA 28.63 USD
46610-056
VWR® Lab Wrap, Clear Plastic
Seals General Purpose Sealing Films
Designed specifically for the lab, clear 100% polyethylene wrap handles easily without tangling, yet seals tightly to a beaker, flask, or Petri dish.
  • Low tangle properties
  • High cling properties
  • Ultra-clean film
Unique formulation yields ultra-clean film to ensure sample purity and eliminate contamination. Dispenser carton features a metal tear-off bar as well as flaps designed to keep the roll in the box.