VWR® Contact Tachometer
Supplier: VWR International
46610-046
46610-046EA 319.36 USD
46610-046
VWR® Contact Tachometer
Tachometers
Unit is designed to measure centrifuges, pumps, rollers, shafts, gears, motors, and moving surfaces.
  • Ensured stable and reproducible results
  • Impact-resistant housing
  • Traceable®
Touch any rotating surface with the cone convex tip, funnel concave tip, or speed wheel for results in 2 seconds. Hold button freezes results. Precision quartz crystal ensures stable and repeatable results. LCD digits are 1.3 cm (1/2") high and can be read from 3m (10') away. Low battery condition is indicated on the display. Impact-resistant ABS housing protects the unit.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with a 9V battery and two tips.