VWR® Enviro-Meter™ Digital Anemometer/Hygrometer/Light Meter/Thermometer
Supplier: VWR International
93676-45
46610-028EA 436.32 USD
46610-028
VWR® Enviro-Meter™ Digital Anemometer/Hygrometer/Light Meter/Thermometer
Anemometers
This meter provides the functions of an anemometer, hygrometer, light meter, and two thermometers in one handheld unit.
  • Four measurements in one unit
  • Lightweight and compact case
The lightweight, compact case permits easy one-handed operation. Bright LCD digital display provides easy-to-read results. Features a hold button that captures a reading for ease in recording a result. Minimum and maximum button displays highest and lowest reading over any time period.
Ordering information: Includes a fast-response, Type K beaded external thermometer probe with 1.2m (4') cable, wrist strap, and 9V battery.