KIMBLE® KONTES® Rotary Evaporator Bump Traps, Fritted, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
166600-0314
46600-834EA 190.64 USD
46600-834 46600-838
KIMBLE® KONTES® Rotary Evaporator Bump Traps, Fritted, DWK Life Sciences
Vacuum Traps
These fritted disc sealed between the lower inner and top outer joint to prevent the contents of the flask from entering into the condenser in the event of bumping.
  • 20 mm diameter
  • 40 to 60 µm porosity fritted disc
  • All traps have a standard taper 24/40 top outer joint
Manufactured from 33 expansion, low extractable borosilicate glass
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.