PTFE-Lined Closed Caps, Thermo Scientific
Supplier: Thermo Scientific
B267-2414UBB
46600-681CS 247.28 USD
46600-681
PTFE-Lined Closed Caps, Thermo Scientific
Closures Bottle Closures
PTFE-Lined Closed Caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials
PTFE-faced silicone liner provides added cushion and an extra secure seal when used with bottles and VOA vials.
The same high-quality PTFE-lined caps used on Thermo Scientific glass environmental sample containers. The PTFE liner is bonded to the cap so it will not fall out or become separated from the cap.