VWR® Non-Return Valves
Supplier: VWR International
46600-040
46600-040CS 364.94 USD
46600-040
VWR® Non-Return Valves
Valves Non Return Valves
This polypropylene non-return valve protects tubing and vacuum systems from back flow when pressure falls in the system.
  • Polypropylene construction
  • Protects against back flow
The recommended working pressure range for this valve is 1.0 to 28.4psi.