Microtiter Tubes, Porex Bio Products Group
Supplier: Thermo Scientific
845
46300-494CS 914.91 USD
46300-494
Microtiter Tubes, Porex Bio Products Group
Tubes Microtubes
Microtiter tubes can be autoclaved, sealed and frozen for storage, and they offer additional volume capacity for sample dilution and/or mixing prior to transfer into microtiter plates
  • Autoclavable
  • Manufactured of superior grade polypropylene
  • Can be frozen to -80 °C ( -112 °F)
Ideal for a wide variety of laboratory procedures, such as HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, blood bank sample freezer storage, and transport of specimens and reagents.