Extractor Kit 1500 Fume Extractor, Nederman
Supplier: Nederman
70511834
46100-570EA 5502.46 USD
46100-570
Extractor Kit 1500 Fume Extractor, Nederman
Fume Extractors
Enables three simultaneous users to capture and filter noxious fumes that occur when soldering, gluing, and working with solvents.
The fan has a variable speed controller that regulates the amount of airflow through the system. Unit is supplied with three extraction arms with an inside diameter of 3.2cm (11/4"). The arms have a 360° swivel at the base and will hold their positions in all directions. Each arm contains a damper to allow for individual users to regulate their own airflow. The clear extraction hood supplied with the kit does not obstruct the view.