PIG® Puncture Repair Kit, New Pig
Supplier: New Pig
PTY203
46000-766EA 219.3 USD
46000-766
PIG® Puncture Repair Kit, New Pig
Spill Control Materials
Contains field-proven tools to stop nearly any drum leak.
  • Includes wooden wedges, pegs, cones, and rubber mallet for fixing larger holes
  • Fix smaller holes with PIG® Repair Putty, washers, screws, lead wool, and tape
A great addition to spill response supplies.