PIG® Spill Containment Tray, New Pig
Supplier: New Pig
PAK371
46000-750EA 68.17 USD
46000-750
PIG® Spill Containment Tray, New Pig
Spill Control Materials
Chemical-resistant polyethylene tray features built-in sump containment to trap leaks and drips while you work.
  • Catch spills when containers leak, spill, or crack
  • Ideal for storing small containers on lab countertop
  • Chemical-resistant polyethylene trays have built-in containment reservoir
  • Translucent sidewalls make leaks visible
  • Pour spout allows easy draining
Translucent sidewalls let you easily inspect fluid level in sump. Compact size fits on tabletop or counter; Excellent for use in labs. Pour spout for easy draining.