PIG® Spill Kit in Small Wall-Mount Cabinet, New Pig
Supplier: New Pig
KIT215
46000-748EA 492.82 USD
46000-748
PIG® Spill Kit in Small Wall-Mount Cabinet, New Pig
Spill Control Materials
Safety-yellow, heavy-gauge, welded steel cabinet is printed "Spill Kit" for quick identification
  • Safety yellow, heavy-gauge, welded steel cabinet is printed "Spill Kit" for quick identification
  • Mat and Pillows absorb; Wipers clean surfaces and hands
Prepacked cabinet provides easy access to contents. See-through front panel lets you easily check inventory levels. Includes write-on label for emergency contact info. Two adjustable shelves.
PIG Mat and Pillows absorb; wipers clean surfaces and hands.