PIG® Elephant Poly-Back Absorbent Mat Roll, New Pig
Supplier: New Pig
MAT270
46000-538EA 399.53 USD
46000-538
PIG® Elephant Poly-Back Absorbent Mat Roll, New Pig
Spill Control Materials
Highly absorbent eight-layer PIG Mat is zigzag stitched to offer excellent walk-on durability.
  • Absorbs and retains oils, coolants, solvents and water
  • Dark gray color hides grime, so mat stays on the job longer
  • Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job
  • Can be wrung out and incinerated after use to reduce waste or for fuels blending
  • Poly backing prevents absorbed liquids from reaching floors and provides added traction for safe footing
  • Rolls are ideal for walkways and large areas to soak up leaks and overspray
Designed specifically for heavy foot traffic or moderate to light wheeled traffic; put the zigzag side up to extend product life. Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Highly absorbent, fine-fiber construction won't leave liquids or fiber residue behind.