Colorsmart Filter for pre-2005 FilterMate Portable Exhausters, Labconco®
Supplier: Labconco
6910201 6910301 6910401 6910501 6933201
46000-236EA 2215 USD
46000-236 46000-238 46000-240 46000-242 46000-244
Colorsmart Filter for pre-2005 FilterMate Portable Exhausters, Labconco®
Filters Gas Filters
Carbon filter for pre-2005 FilterMate portable exhausters.
Contain a minimum of 26 pounds of impregnated carbon. Color-coded. Includes three sampling tubes that detect a broad range of chemicals specific to the filter. For use as replacement filter in discontinued Protector FilterMate Mobile Stations currently in service.