Anticon® Gold HeavyWeight™ Wipes, Contec®
Supplier: Contec
492224-798 492224-771 492224-789 492224-792 492224-790 492224-783
45800-054CS 1019.96 USD
45800-054 45800-058 45800-060 45800-062 45800-064 45800-134
Anticon® Gold HeavyWeight™ Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Anticon® Gold HeavyWeight™ wipes are made from 100% continuous filament polyester with a double-knit, no-run interlock construction. They use patented textile technology to achieve superior sorption characteristics for critical applications.
  • Sealed edges to reduce particle and fiber generation
  • High sorptive capacity and fast adsorption rate for superior spill control
  • High tensile strength eliminates tearing and snagging in tough cleaning applications
  • Specially processed for superior cleanliness and sorbency
The proprietary sealed edges of these wipes allows the use of 100% of the wipe for cleaning without the worry of harming delicate surfaces. Anticon® Gold HeavyWeight™ wipes' low sub-micron particles and fiber counts are ideal for use in the most critical environments. Cleanroom laundered and packaged. Also available with Particle Attraction Technology (P.A.T.).
Certifications: Recommended for ISO Class 3 to 5 environments.