Anticon® Sterile Gold Cleanroom Wiper, Contec®
Supplier: Contec
492226-942
45800-038CS 638.22 USD
45800-038
Anticon® Sterile Gold Cleanroom Wiper, Contec®
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
Anticon® Sterile Gold™ Wipes are a sterile version of Anticon® Gold HeavyWeight™ wipes. These wipes are made from 100% continuous filament polyester with a double-knit, no-run interlock construction. The proprietary sealed edges reduces the amount of fibres released from the wipe.
  • Patented textile technology creates a polyester wipe with high sorbency
  • High sorptive capacity decreases the number of wipes required for standard use
  • Excellent choice for wiping and absorbing spills in critical cleanroom environments
  • Packed in small quantities in easy tear bags to reduce waste
Wipes are cleanroom laundered and packed in a class 4 cleanroom. They exhibit very low particle and fiber counts, which helps to minimize contamination. These wipes are ideal for use in the most critical of cleanroom environments. Using patented textile technology, they achieve superior capacity with a quick sorption rate. Ideal for disinfectant and solvent application and wiping to dry.
Anticon® Sterile Gold™ wipes are ideal for use in the most critical aseptic environments. Wipes are sterilized via gamma irradiation to a sterility assurance limit of 10⁻⁶ and a certificate of irradiation is provided with each shipment.
Certifications: Recommended for ISO Class 5 environments.
Packaging: They are conveniently packaged in easy tear bags containing only 20 wipes per package to eliminate waste.