Anticon® Sterile 100™ Wipes, Contec®
Supplier: Contec
492226-939
45800-032CS 805.97 USD
45800-032
Anticon® Sterile 100™ Wipes, Contec®
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
Anticon® Sterile 100™ are the sterile version of Anticon 100® HeavyWeight™ wipes. They are made from 100% continuous filament polyester with a double-knit, no-run interlock construction. The clean cut edges create a very soft wipe, where the whole of the wipe fabric can be used with no risk of scratching.
  • Recommended for ISO Class 5 environments
  • Validated sterile for aseptic environments
  • Patented textile technology creates a polyester wipe with high sorbency
  • Lowest fibers and particles available for a cut edge wipe reduces risk of contamination
  • High sorptive capacity decreases the number of wipes required for standard use
  • Excellent choice for wiping and absorbing spills in critical cleanroom environments
Wipes are cleanroom laundered and packed in a class 4 cleanroom. They exhibit very low particle and fiber counts for a knife cut wipe, which helps to minimize contamination. Using patented textile technology, these wipes achieve superior capacity with a quick sorption rate, and are ideal for use in critical pharmaceutical environments. They are also excellent for disinfectant and solvent application and wiping to dry.
Anticon® Sterile 100™ wipes are conveniently packaged in easy tear bags containing 20 or 25 wipes per package. They are sterilized via gamma irradiation to a sterility assurance level of 10⁻⁶ and a certificate of irradiation is provided with each shipment. These wipes are the same substrate as Anticon® Gold HeavyWeight™ wipes, but with cut edges.