Quartz SUPRASIL Macro Cell with Inlet/Outlet Tubes on Top, PerkinElmer
Supplier: PerkinElmer
B0631126
45603-882EA 1369.46 USD
45603-882
Quartz SUPRASIL Macro Cell with Inlet/Outlet Tubes on Top, PerkinElmer
Cuvettes Spectrophotometer Cuvettes
Fluorescence flow-through Cells with macro inlet/outlet tubes on top for measuring samples with continuous flow.
Flow-through cells are used for measuring samples with continuous flow or with sippers where individual samples are aspirated into the cell. All cells have center heights of 15 mm. Each package contains one cell with two clear windows. The cells listed below are compact with two screw connectors, M6×1 and 500 mm length FEP tubing - 1.9 mm OØ, 1.1 mm IØ.