Special Optical Glass Macro Cylindrical Cell with PTFE Stopper, PerkinElmer
Supplier: PerkinElmer
B0631094
45603-834EA 228.87 USD
45603-834
Special Optical Glass Macro Cylindrical Cell with PTFE Stopper, PerkinElmer
Cuvettes Spectrophotometer Cuvettes
The special optical glass macro cylindrical cell has a volume of 28 ml, a light path of 100 mm, and an outside depth of 102.5 mm.
Cylindrical cells are used with spectrophotometers that have a larger circular light beam. Each cell has an outside diameter of 22 mm and an inside diameter of 19 mm. Each package contains one cell and two windows.