Quartz SUPRASIL® Macro and Micro Cells, PerkinElmer
Supplier: PerkinElmer
B0631029 B0631009 B0631077 B0631011 B0631027 B0631079 B0631071 B0631025 B0631008 B0631012 B0631049 B0631007 B0631026 B0631030 B0631080 B0631083 B0631010
45603-742EA 1120.74 USD
45603-742 45603-810 97012-870 97012-874 97012-878 97012-882 97012-884 97012-924 97013-524 97013-536 97013-544 97013-546 45603-698 45603-736 45603-740 45603-744 45603-788 45603-808 45603-812 45603-814 97012-872
Quartz SUPRASIL® Macro and Micro Cells, PerkinElmer
Cuvettes Spectrophotometer Cuvettes
Standard rectangular macro and micro cells are the most frequently used type of spectroscopy cell for routine liquids analysis.
  • Clear windows for easy observation
  • Widely used in spectroscopy procedures
  • Rectangular design
Quartz SUPRASIL® cells are available in a range of sizes for use with various sample volumes. All cells contain two clear windows.