Integrated Platform THGA Graphite Tubes, PerkinElmer
Supplier: PerkinElmer
B0504033 B3000655 B3000641 B3000653 N3110147
97013-424PK 7535.6 USD
97013-424 97013-428 97013-430 97013-432 45603-448
Integrated Platform THGA Graphite Tubes, PerkinElmer
Tubes Graphite Tubes
These transversely-heated graphite tubes are made from a blend of high-quality base graphite material, providing the user with the required performance to yield reproducible results from day to day with respect to sensitivity (characteristic mass), accuracy and precision.
  • Integrated platform provides an even temperature heating surface
  • Significantly reduces or eliminates condensation of the sample matrix components and 'memory' effects
  • Improves the atomization efficiency for refractory elements
The graphite tube, the heart of the graphite furnace, plays a vital role in the overall stability of an analysis. In order to assure stable analytical conditions from one atomization cycle to the next, and from graphite tube to graphite tube, all graphite parts - contacts, tubes and platforms - must be subject to strict quality control by both the instrument manufacturer and the graphite manufacturer.
For improved characteristic mass with volatile and medium refractory elements such as Ag, As, Cd, Co, Cr, Cu, Pb, Se, and Tl, the THGA graphite tubes with end-caps are recommended. Additional benefits of these tubes include: Improved detection limits for volatile and medium refractory elements and Faster drying for complex matrices due to enlarged dosing hole.
THGA graphite tubes with end caps are not recommended for refractory elements such as V or for Ba.
The tubes are compatible with PerkinElmer PinAAcle 900T, PinAAcle 900Z, AAnalyst 600, AAnalyst 800, Model 4100ZL, Model 4110ZL, Model 5100ZL, SIMAA 6000 Zeeman AA instruments.