Zinc (Zn) System 2 Electrodeless Discharge Lamp, PerkinElmer
Supplier: PerkinElmer
N3050691
45603-442EA 1626.14 USD
45603-442
Zinc (Zn) System 2 Electrodeless Discharge Lamp, PerkinElmer
Lights Hollow Cathode Lamps
System 2 Electrodeless Discharge Lamps (EDLs) provide the optimized spectral output needed to get the maximum performance from PerkinElmer® Atomic Absorption (AA) spectrometers.
  • System 2 Electrodeless Discharge Lamp for the detection of elemental Zinc (Zn)
  • PinAAcle™ instruments require the purchase of an EDL driver assembly for each of the lamp positions to be used
Electrodeless Discharge Lamps for greater brightness (EDLs) are typically much brighter and, in some cases, provide better sensitivity than comparable HCLs. EDLs are preferred for certain volatile elements. They offer better precision and lower detection limits for analyses that are “noisy” due to weak hollow cathode emission. Electrodeless Discharge Lamps (EDLs) are designed to be physically interchangeable with Intensitron Hollow Cathode Lamps. Their power supplies are built into all the PinAAcle™ series of atomic absorption spectrometers.