Bismuth (Bi) System 2 Electrodeless Discharge Lamp, PerkinElmer
Supplier: PerkinElmer
N3050611
45603-416EA 1626.14 USD
45603-416
Bismuth (Bi) System 2 Electrodeless Discharge Lamp, PerkinElmer
Lights Hollow Cathode Lamps
System 2 electrodeless discharge lamp for the detection of elemental Bismuth (Bi). System 2 electrodeless discharge lamps (EDLs) provide the optimized spectral output needed to get the maximum performance from PerkinElmer® atomic absorption spectrometers.
Note: PinAAcle™ instruments require the purchase of an EDL driver assembly (97013-414), for each of the lamp positions to be used.
Electrodeless discharge lamps for greater brightness (EDLs) are typically much brighter and, in some cases, provide better sensitivity than comparable HCLs. EDLs are preferred for certain volatile elements.More Product Information