Praseodymium (Pr) Lumina™ Hollow Cathode Lamp, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N3050161
45603-330EA 2045.6 USD
45603-330
Praseodymium (Pr) Lumina™ Hollow Cathode Lamp, PerkinElmer
Lights Hollow Cathode Lamps
Single Element Lumina™ hollow cathode lamp (HCL) for the detection of elemental Praseodymium (Pr). Series N30501XX Lumina™ 2" (50 mm) diameter single-element lamps are designed to be used with our PinAAcle™ and AAnalyst™ atomic absorption spectrometer series of instruments.
Please note that this element requires the use of a nitrous oxide-acetylene flame. Every genuine , PerkinElmer lamp is designed for use with and tested on our spectrometers to assure compatibility and the highest performance. We offer a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ Hollow Cathode Lamp (HCL), we produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume - and the longer the lamp lifetime. Note: Lumina™ Hollow Cathode Lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable.