PEEK Tubing, PerkinElmer
Supplier: PerkinElmer
N9302650
45600-118EA 152.49 USD
45600-118
PEEK Tubing, PerkinElmer
Tubing Chromatography Tubing
PEEK tubing has the strength required to withstand continuous use at HPLC pressure without swelling or bursting.
  • Can be used virtually with any organic or inorganic liquid
  • Not affected by halide salts, high-strength buffers, or other aggressive mobile phases
PEEK tubing acts as an excellent altnerative for stainless steel tubing in high-pressure applications and analytical systems. Its mechanical properties allow it to be used in many situations where stainless steel would become too reactive.