Hydrosep® Eye Wash Bottle, Encon
Supplier: Encon Safety Products
01110764
45500-180CS 173.46 USD
45500-180
Hydrosep® Eye Wash Bottle, Encon
First Aid Supplies Eyewash Stations and Eyewash Bottles
Recommended cleaning and fluid replacement every 90 to 120 days in unsealed, gravity-fed eyewashes.
  • Duration of effectiveness is dependent upon type of device
  • Proven effective for 180 days in a sealed, pressurized, portable, or air-driven TWS shower system