Corning® TE Buffer, 1X, Liquid, Corning
Supplier: Corning
46-009-CM
45001-155PK 416.88 USD
45001-155
Corning® TE Buffer, 1X, Liquid, Corning
Buffers Cloning Buffers
Tris-EDTA (TE) buffer is commonly used in work with nucleic acids.
Examples include as a solubilization buffer in DNA and RNA purification, as a running buffer in electrophoresis, and as a general storage buffer for nucleic acids.