Sodium chloride solution 5M DNase-, RNase-, protease-free, Corning®

Supplier: Corning

46-032-CV
45001-125PK 372.88 USD
45001-125
Sodium chloride solution 5M DNase-, RNase-, protease-free, Corning®
Sodium chloride

The 5 M NaCl is a common molecular biology reagent.


All production lots are tested to ensure that they are free of RNAse. This product is used to control osmolarity and conductivity, among other factors.

Formula: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR