Dimethyl sulfoxide, sterile filtered

Supplier: Corning
Warning

25-950-CQC
45001-118EA 134.19 USD
45001-118
Dimethyl sulfoxide, sterile filtered
Dimethyl sulphoxide

Dimethyl Sulfoxide (DMSO) is commonly used in cell cryopreservation, along with glycerol, to prevent damage to the cell membrane during freezing.


  • Endotoxin tested
  • Septum cap
  • Shelf life 12 months


Mediatech, lnc. products are prepared by a validated aseptic sterile filtration process to ensure that ali products comply with the industry's Sterility Assurance Level (SAL) of at least 1 o-3 (i.e. demonstrating th at a product produced in this manner has no more th an 1 random contaminant per 1000 units).
This product was tested in accordance with currently approved Mediatech, lnc. specifications and procedures. Testing procedures are maintained in compliance with the current versions of the USP and/or EP , where applicable.
For Research Use Only. Not for Human Therapeutic Use. Utilization of this product apart from the labeled intended use may be a violation of local and/or Federal Law.

Formula: (CH₃)₂SO
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 18.5 °C
Density: 1.101 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR