D-(+)-Glucose solution 45%, Corning®

Supplier: Corning

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

25-037-CI
45001-116EA 46.05 USD
45001-116
D-(+)-Glucose solution 45%, Corning®
D(+)-Glucose

45% Glucose Solution. Glucose is a main source of energy for living organisms and is used as a nutrient source in cell culture media. This 45% (w/v) solution has been to ensure no cytotoxicity.

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00148912
CAS Number: 50-99-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

pH 3.5 ± 0.5 @ 20-25°
Osmolality 250-280 mOsm/Kg H20 @ 1:10
Endotoxin ≤ 0.25 EU/ml @ FW
Sterility Pass
Cytotoxicity Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR