Corning® MCDB 131 1X, Corning
Supplier: Corning
15-100-CV
45001-112PK 353.95 USD
45001-112
Corning® MCDB 131 1X, Corning
Cell Culture Media
MCDB 131 was originally developed by the researchers Knedler and Ham for the low-protein and serum free growth of human microvascular endothelial cells (HMVEC)
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
Since its development, this medium has been used for this cell type and others such as liver, smooth muscle, cardiac myocytes. It is one of a group of MCDB media which were designed to provide a defined and optimally balanced nutritional environment for various mammalian cell types.