Penicillin/Streptomycin/Glutamine 100X, Corning®
Supplier: Corning
30-009-CI
45001-103PK 259.93 USD
45001-103
Penicillin/Streptomycin/Glutamine 100X, Corning®
Penicillin/Streptomycin/Glutamine
Penicillin-Streptomycin-L-glutamine combines two antibiotics with a nutrient which is essential for cell growth, L-glutamine
  • 10,000 I.U. Penicillin (per mL)
  • 10,000 ug/mL Streptomycin
  • 29.2 mg/mL L-glutamine
L-Glutamine is essential as an energy source for most mammalian cells in culture because of its amino acid carbon skeleton.
This antibiotic-nutrient combination is effective against Gram-negative and Gram-positive bacteria.
Penicillin inhibits bacterial growth by inhibiting peptidoglycan synthesis (essential for the bacterial cell wall), and streptomycin inhibits bacterial growth by inhibiting protein synthesis.