Carbenicillin disodium salt, powder

Supplier: Corning

Synonyms: NSC 111071, α-Carboxybenzylpenicillin disodium salt, Sodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na

46-100-RG
45001-094EA 397.41 USD
45001-094
Carbenicillin disodium salt, powder
Carbenicillin disodium salt

Carbenicillin is a form of penicillin and is often used in place of ampicillin because it maintains stability after being autoclaved.


  • Can be autoclaved, unlike ampicillin


Like ampicillin, carbenicillin interferes with cell-wall synthesis of actively growing cells.

Formula: C₁₇H₁₆N₂O₆S·2Na
MW: 422.363 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077683
CAS Number: 4800-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR