X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside), powder, Corning®

Supplier: Corning

46-101-RF
45001-092EA 380.57 USD
45001-092
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside), powder, Corning®
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)

X-Gal (5-bromo-4-chloro-3-indolyl-b-D-galactopyranoside) is a substrate of b-galactosidase, a gene product of the lac operon in E. coli.


X-Gal included in bacterial medium will be hydrolyzed by ß-galactosidase to produce indoxyl, a colormetric indicator of the activity of the lac operon.

Formula: C₁₄H₁₅BrClNO₆
Melting Pt: 237…239 °C
MDL Number: MFCD00005666
CAS Number: 7240-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR