Corning® Insulin-Transferrin-Selenium (ITS) 100x Growth Supplement, Corning
Supplier: Corning
25-800-CR
45001-090EA 69.75 USD
45001-090
Corning® Insulin-Transferrin-Selenium (ITS) 100x Growth Supplement, Corning
Cell Culture Supplements and Additives
Animal free formulation
Contains:
1000 mg/L insulin, human recombinant
550 mg/L transferrin, human recombinant
0.67 mg/L selenious acid
Caution: Storage 2°C to 8°C. For research use only.