Corning® glutagro™ Supplement, Corning
Supplier: Corning
25-015-CI
45001-086EA
60.39
USD
45001-086
Corning® glutagro™ Supplement, Corning
Cell Culture Supplements and Additives
Cells grown in traditional media supplemented with L-glutamine are susceptible to toxicity
With Corning® glutagro™, cells receive this needed supplement in the healthiest environment possible. This modified form of L-glutamine does not degrade into ammonia in storage and incubation, minimizing toxicity and improving culture quality.