Corning® 10X TAE Buffer, Liquid, Corning
Supplier: Corning
46-010-CM
45001-074PK 339.36 USD
45001-074
Corning® 10X TAE Buffer, Liquid, Corning
Buffers Electrophoresis Buffers Running Buffers
Tris-Acetate-EDTA (TAE) buffer is used in DNA agarose gel electrophoresis, both in the agarose gel itself and the running buffer
Linear, double-stranded DNA separates faster in a TAE buffer-based system compared to Tris-Borate-EDTAA (TBE) buffer, but the latter has a higher buffering capacity and is thus more resistant to changes in pH. Use of TAE vs. TBE buffer depends on downstream applications. TAE is recommended for preparative electrophoresis in which the nucleic acid bands being separated are to be used in cloning, and other work requiring enzymatic applications.