Corning® insectagro® Serum-Free/Protein-Free Medium, 1X, Corning
Supplier: Corning
13-410-CV
45001-054PK
316.7
USD
45001-054
Corning® insectagro® Serum-Free/Protein-Free Medium, 1X, Corning
Cell Culture Media
Corning® insectagro® Sf9 is formulated to support the propagation of Sf9 insect cells in culture, and can also be used for SF21 cells
Sf9 cells are cultured in non-humidified, non-CO2 incubators at 27ºC (room temperature) and display both monolayer and suspension culture qualities. With their fast doubling times of 10-22 hours, Sf9 cells are easily scaled up to large cultures using bioreators.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)