Corning® Terrific Broth, Corning
Supplier: Corning
46-055-CM
45001-050PK 392.83 USD
45001-050
Corning® Terrific Broth, Corning
Media for Microbiology Dehydrated Media for Microbiology
Terrific Broth is a medium for the propagation and maintenance of recombinant E.coli.
Because this medium supports higher cell densities than LB medium, it is preferred for maximizing plasmid yield in preparation for nucleic acid purification.