Corning® Miller's LB Broth, Corning
Supplier: Corning
46-050-CM
45001-048PK 366.37 USD
45001-048
Corning® Miller's LB Broth, Corning
Media for Microbiology Dehydrated Media for Microbiology
Miller's LB broth is based on the original Lysogeny Broth (LB) media formulation developed by Luria and Bertani, which was designed for the rapid propagation of recombinant E. coli strains.
  • 20-fold concentration sodium chloride compared to the original formulation
  • Widely used for the growth and maintenance of cells
  • Work with plasmid DNA and recombinant protein production
Delivery information: Contents: Casein peptone (10 g/L), UF Yeast Powder (5 g/L), NaCl (10 g/L)