Amphotericin B 250 µg/ml

Supplier: Corning

30-003-CF
45000-808PK 223.51 USD
45000-808
Amphotericin B 250 µg/ml
Amphotericin B

This antimycotic interferes with fungal membrane permeability by forming channels in cell membranes, causing small molecules to leak from the cell. It is recommended for use in cell culture applications at 10 ml/L and is stable at 37 ºC for 3 days.


Packaging: Shipped frozen.

Formula: C₄₇H₇₃NO₁₇
Storage Temperature: Freezer
MDL Number: MFCD00877763
CAS Number: 1397-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR