Hygromycin B solution 50 mg/ml, sterile

Supplier: Corning

Synonyms: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol

30-240-CR
45000-806EA 289.71 USD
45000-806
Hygromycin B solution 50 mg/ml, sterile
Hygromycin B

Hygromycin B, produced by Streptomyces hygroscopicus, is used as a selection agent and inhibits protein synthesis in cells not carrying hygromycin phosphotransferase (HPH)


Antibiotic; For selection/maintenance of prokaryotic, eukaryotic cells.


Application: Recommended for use as a selection agent at 100-800 μg/mL. Biochem/physiol Actions: Mode of Action: Blocks polypeptide synthesis and inhibits elongation. For use in the selection and maintenance of prokaryotic and eukaryotic cells. Preparation Note: Purified by ion exchange chromatography.


HPH inactivates hygromycin B and restores protein synthesis.


Packaging: Shipping temperature: Ambient

Formula: C₂₀H₃₇N₃O₁₃
MW: 527.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06795479
CAS Number: 31282-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR