Corning® Polysucrose 400, Corning
Supplier: Corning
Polysucrose 400 is a synthetic polymer of sucrose and epichlorohydrin used for the density gradient separation of cells, cell membranes and virus cells
It can also be used to isolate different organelles. This nonionic compound is provided in powder form.