Sodium pyruvate solution 100 mM with 8.5 g/L NaCl cell culture reagent, Corning®

Supplier: Corning

25-000-CI
45000-710PK 95.68 USD
45000-710
Sodium pyruvate solution 100 mM with 8.5 g/L NaCl cell culture reagent, Corning®
Sodium pyruvate

100 mM Solution with 8.5 g/L NaCl Sodium pyruvate is often used as a component in culture broth and media as an energy source for propagating cells. This reagent is also used in Wallen fermentation medium to enhance the conversion of oleic acid to 10-ketostearic acid.


It is provided as a 100 mM Solution with 8.5 g/L NaCl.

Formula: CH₃COCOONa
Storage Temperature: Refrigerator
MDL Number: MFCD00002586
CAS Number: 113-24-6

Order Now

Specification Test Results

pH 6.0 ± 1.0
Osmolality 400 ± 100 mOsm/Kg H20
Endotoxin < 0.25 EU/ml@ 1:100
Mycoplasma Tested Negative
Sterility Pass
Cell Growth Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR