Corning® MEM (Minimum Essential Medium) Nonessential Amino Acids
Supplier: Corning
25-025-CI
45000-700PK 156 USD
45000-700
Corning® MEM (Minimum Essential Medium) Nonessential Amino Acids
Cell Culture Supplements and Additives
The addition of MEM nonessential amino acids to medium can improve growth and viability of propagating cells in culture while reducing the biosynthetic burden on these cells
This concentrated supplement allows for the efficient addition to media with no risk of dilution of media components.