L(+)-Glutamine solution 200 mM (100x) in water + 8.5 g/L NaCl cell culture reagent, Corning®

Supplier: Corning

25-005-CV
45000-678PK 717.95 USD
45000-678
L(+)-Glutamine solution 200 mM (100x) in water + 8.5 g/L NaCl cell culture reagent, Corning®
L(+)-Glutamine

L-glutamine is an essential amino acid and a key component of culture media, serving as a major energy source for propagating cells. It is very stable as a dry powder and as a frozen solution but degrades rapidly in liquid media or stock solutions, producing toxic compounds. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use. This formulation is prepared in cell culture grade water.


Caution: Storage -25°C to -15°C

Formula: C₅H₁₀N₂O₃
MDL Number: MFCD00008044
CAS Number: 56-85-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb7a812ec5d2c73a797fc464926df9eef12c5c9a&vid=15d7d87daff0fec33a974a578415e157a3278049&prid=web&clid=SAPDAM

Order Now

Specification Test Results

pH 5.5 ± 0.5
Osmolality 450 ± 50 mOsm/kg H2O
Endotoxin < 0.25 EU/ml @ 1 :50
Mycoplasma Tested Negative
Sterility Pass
Cell Growth Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR