Corning® Cellstripper® Liquid, Corning
Supplier: Corning
25-056-CI
45000-668PK 130.58 USD
45000-668
Corning® Cellstripper® Liquid, Corning
Cell Culture Supplements and Additives
Cellstripper® is a non-enzymatic cell dissociation solution formulated with a proprietary mixture of chelators that gently dislodge adherent cells in tissue culture. It is an alternative to trypsin when used with either serum-free or serum-supplemented media. Because Cellstripper® is a gentler reagent than trypsin, cells can be exposed to Cellstripper for longer periods of time. Cellstripper® achieves superior results in comparison to enzymatic dissociation products.