Corning® Trypsin
Supplier: Corning
25-054-CI
45000-666PK 252.23 USD
45000-666
Corning® Trypsin
Enzymes
Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging
  • Trypsin 10X 2,5% Trypsin in HBSS without calcium, magnesium, and phenol red Porcine Parvovirus Tested
Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action. EDTA sequesters these ions and thus enhances the efficacy of trypsin.