Corning® Trypsin
Supplier: Corning
25-050-CI
45000-658PK 161.06 USD
45000-658
Corning® Trypsin
Enzymes
0.25% Trypsin in HBSS (Hank’s Balanced Salt Solution) without calcium and magnesium.
  • Porcine parvovirus tested
  • 6×100 ml
Trypsin is an enzymatic reagent that is used to release adherent cells from tissue plates for passaging. This offering contains trypsin at a concentration of 0.25%.