Tetracycline hydrochloride, powder

Supplier: Corning

61-242-RG
45000-656EA 100.49 USD
45000-656
Tetracycline hydrochloride, powder
Tetracycline hydrochloride

Tetracycline Hydrochloride (HCl) is an antibiotic that inhibits protein synthesis by binding to the 30S ribosomal subunit, blocking interactions at the acceptor site on the RBA-ribosome complex


It is a member of the tetracycline group of antibiotics, a broad-spectrum class that inhibits growth of both aerobic and anaerobic organisms through this common mechanism. The congener tetracycline is semi-synthetically produced from chlortetracycline, a product of Staphylococcus aureofaciens. Resistance is conferred by the TetR-Tn10 gene product that blocks cell wall permeability. Tetracycline HCl is effective against Gram-positive and -negative bacteria and is offered in powder form.


Packaging: Shipping temperature: Ambient

Formula: C₂₂H₂₅ClN₂O₈
MW: 480.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078142
CAS Number: 64-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR